missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-azaindole, 97%, Thermo Scientific™

Product Code. 15585206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15585206 Supplier Thermo Scientific Acros Supplier No. 456710010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 55052-28-3
Melting Point 176.0°C to 181.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H5ClN2
MDL Number MFCD08272232
Quantity 1 g
Synonym 4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine
InChI Key HNTZVGMWXCFCTA-UHFFFAOYSA-N
SMILES ClC1=C2C=CNC2=NC=C1
IUPAC Name 4-chloro-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol) 152.58
PubChem CID 11389493
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 4-Chloro-7-azaindole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-7-azaindole, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.