missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-azaindole, TRC

Product Code. 30368428
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368428 1 g 1g
30361213 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368428 Supplier Toronto Research Chemicals Supplier No. TRCC3645001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55052-28-3
Molecular Formula C7 H5 Cl N2
Molecular Weight (g/mol) 152.58
IUPAC Name 4-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES Clc1ccnc2[nH]ccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5ClN2/c8-6-2-4-10-7-5(6)1-3-9-7/h1-4H,(H,9,10)
Formula Weight 152.0141
Chemical Name or Material 4-Chloro-7-azaindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.