missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-8-(trifluoromethyl)quinoline, TRC

Product Code. 30365758
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30365758 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30365758

Brand: Toronto Research Chemicals TRCC4220505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-8-(trifluoromethyl)quinoline
CAS 23779-97-7
Molecular Formula C10H5ClF3N
Quantity 5 g
Synonym 4-Chloro-8-(trifluoromethyl)-quinoline
InChI Formula InChI=1S/C10H5ClF3N/c11-8-4-5-15-9-6(8)2-1-3-7(9)10(12,13)14/h1-5H
SMILES FC(F)(F)c1cccc2c(Cl)ccnc12
IUPAC Name 4-chloro-8-(trifluoromethyl)quinoline
Molecular Weight (g/mol) 231.6
Formula Weight 231.0063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.