missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-toluenesulfonyl chloride, 97%

Product Code. 11391929
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391929 5 g 5g
11381929 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391929 Supplier Thermo Scientific Alfa Aesar Supplier No. H26017.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6966-45-6 | C7H6Cl2O2S | 225.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6966-45-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD01631927
InChI Key DBJRPJSDYFDWPV-UHFFFAOYSA-N
Synonym 4-chlorophenyl methanesulfonyl chloride, 4-chlorobenzylsulfonyl chloride, 4-chloro-phenyl-methanesulfonyl chloride, 4-chloro-alpha-toluenesulfonyl chloride, chloro 4-chlorophenyl methyl sulfone, 4-chlorobenzenemethanesulfonyl chloride, 4-chlorophenyl methanesulphonyl chloride, 4-chlorophenyl methyl sulfonyl chloride, acmc-20apnh
PubChem CID 227304
IUPAC Name (4-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)Cl

Specifications

Melting Point 93°C to 96°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 225.1
Percent Purity 97%
Chemical Name or Material 4-Chloro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.