missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-m-phenylenediamine, 97%

Product Code. 11489150
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489150 5 g 5g
11499150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489150 Supplier Thermo Scientific Alfa Aesar Supplier No. A15430.06
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5131-60-2 | C6H7ClN2 | 142.586 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5131-60-2
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00025284
InChI Key ZWUBBMDHSZDNTA-UHFFFAOYSA-N
Synonym 4-chloro-m-phenylenediamine, 1,3-benzenediamine, 4-chloro, 4-chloro-1,3-benzenediamine, 4-chloro-1,3-phenylenediamine, 1-chloro-2,4-diaminobenzene, 4-chloro-meta-phenylenediamine, m-phenylenediamine, 4-chloro, 4-chlorophene-1,3-diamine, 4-chloro-1,3-diaminobenzene, 4-chlorophenylene-1,3-diamine
PubChem CID 21209
ChEBI CHEBI:82421
IUPAC Name 4-chlorobenzene-1,3-diamine
SMILES C1=CC(=C(C=C1N)N)Cl

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-m-phenylenediamine
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-m-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Chloro-m-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.