missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59572.03

Code : NEW

Additional Details : CAS Number : 591242-96-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475938

  • 646.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

591242-96-5
263.696
PGDMFVGTVYSLDE-UHFFFAOYSA-N
958087
CC1=C(C=CC(=C1)F)NC(=O)C2=CC=C(C=C2)Cl
C14H11ClFNO
MFCD04084666
4-chloro-n-4-fluoro-2-methylphenyl benzamide
4-chloro-N-(4-fluoro-2-methylphenyl)benzamide

Specifications

4-Chloro-N-(4-fluoro-2-methylphenyl)benzamide
591242-96-5
MFCD04084666
PGDMFVGTVYSLDE-UHFFFAOYSA-N
4-chloro-N-(4-fluoro-2-methylphenyl)benzamide
958087
Solid
1g
C14H11ClFNO
4-chloro-n-4-fluoro-2-methylphenyl benzamide
CC1=C(C=CC(=C1)F)NC(=O)C2=CC=C(C=C2)Cl
263.696
263.7
97%
Videos
SDS