missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-phenylenediamine, 97%

Product Code. 11469772
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469772 500 g 500g
11449772 25 g 25g
11459772 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469772 Supplier Thermo Scientific Alfa Aesar Supplier No. A10823.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N

Spezifikation

Melting Point 70°C to 74°C
Quantity 500 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-o-phenylenediamine

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.