missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloroacetyl)morpholine, TRC

Product Code. 30356870
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356870 10 g 10g
30355451 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30356870 Supplier Toronto Research Chemicals Supplier No. TRCC36370010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1440-61-5
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.6
Synonym 2-Chloro-1-(4-morpholinyl)ehanone, 2-Chloro-1-(morpholin-4-yl)ethanone, 2-Chloro-1-morpholino-1-ethanone, 2-Chloro-1-morpholinoethanone, 4-Morpholinylcarbonylmethyl Chloride, Chloroacetic Acid Morpholide, Chloroacetic Acid Morpholine Amide, N-(Chloroacetyl)morpholine, NSC 54542
IUPAC Name 2-chloro-1-morpholin-4-ylethanone
SMILES ClCC(=O)N1CCOCC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H10ClNO2/c7-5-6(9)8-1-3-10-4-2-8/h1-5H2
Formula Weight 163.04
Chemical Name or Material 4-(Chloroacetyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.