missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline-2,6-d2, TRC

Product Code. 30368663
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368663 25 mg 25mg
30358440 2.5 mg 2.5mg
30361168 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30368663 Supplier Toronto Research Chemicals Supplier No. TRCC37750125MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35749-94-1
Molecular Formula C6H4D2ClN
Molecular Weight (g/mol) 129.58
Synonym 4-Chloroaniline-2,6-D2, 4-Chloroaniline D2 (phenyl 2,6 D2), Benzen-2,6-d2-amine, 4-chloro-, 4-Chloroaniline-2,6-d2, 4-Chlorobenzen-2,6-d2-amine
IUPAC Name 4-chlorobenzen-2,6-d2-amine
SMILES ClC1=CC([2H])=C(C([2H])=C1)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2/i3D,4D
Formula Weight 129.03
Chemical Name or Material 4-Chloroaniline-2,6-D2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.