missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline (4-Chlorophenylamine), TRC

Product Code. 30134903
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134903 5 g 5g
30040681 1 g 1g
30040579 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134903 Supplier Toronto Research Chemicals Supplier No. TRCC3775005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-47-8
Molecular Formula C6 H6 Cl N
Molecular Weight (g/mol) 127.57
Synonym Aniline, p-chloro- (8CI), 4-Chlorobenzenamine, 1-Amino-4-chlorobenzene, 4-Amino-1-chlorobenzene, 4-Aminochlorobenzene, 4-Chloro-1-aminobenzene, 4-Chloroaniline, 4-Chlorophenylamine, NSC 36941, p-Aminochlorobenzene, p-Chloroaniline, p-Chlorophenylamine
IUPAC Name 4-chloroaniline
SMILES Nc1ccc(Cl)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2
Formula Weight 127.0189
Percent Purity >95%
Chemical Name or Material 4-Chloroaniline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.