missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline, Extra Pure, SLR, Fisher Chemical

Product Code. 10630331 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10630331 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10630331 Supplier Fisher Chemical Supplier No. C/4600/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 68°C
CAS 106-47-8
Color Brown
Quantity 100 g
Packaging Plastic powder jar
Physical Form Solid
Molecular Formula C6H6ClN
MDL Number 7835
Boiling Point 232°C
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES NC1=CC=C(Cl)C=C1
IUPAC Name 4-chloroaniline
Molecular Weight (g/mol) 127.57
Formula Weight 127.57g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.