missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile, 99%

Product Code. 11471878
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471878 50 g 50g
11481878 250 g 250g
11461878 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471878 Supplier Thermo Scientific Alfa Aesar Supplier No. A14524.18
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-03-0 | C7H4ClN | 137.57 g/mol

4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-03-0
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001813
InChI Key GJNGXPDXRVXSEH-UHFFFAOYSA-N
Synonym p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
PubChem CID 12163
IUPAC Name 4-chlorobenzonitrile
SMILES ClC1=CC=C(C=C1)C#N

Specifications

Melting Point 90°C to 94°C
Density 1.2
Quantity 50 g
Odor Almond-like
Beilstein 1072122
Boiling Point 222°C to 224°C
Solubility Information Soluble in alcohol and ether. Insoluble in water.
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 4-Chlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.