missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl mercaptan, 98%, Thermo Scientific™

Product Code. 11354749
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11354749

Brand: Thermo Scientific Alfa Aesar A19876.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6258-66-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00004870
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
PubChem CID 80409
IUPAC Name (4-chlorophenyl)methanethiol
SMILES SCC1=CC=C(Cl)C=C1

Specifications

Melting Point 19°C to 20°C
Density 1.202
Boiling Point 234°C
Quantity 5 g
Odor Stench
Refractive Index 1.5895
UN Number UN2810
Beilstein 606496
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzyl mercaptan
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.