missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl mercaptan, 98%, Thermo Scientific™

Product Code. 11478707
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478707 25 g 25g
11354749 5 g 5g
11488707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478707 Supplier Thermo Scientific Alfa Aesar Supplier No. A19876.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6258-66-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00004870
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
PubChem CID 80409
IUPAC Name (4-chlorophenyl)methanethiol
SMILES SCC1=CC=C(Cl)C=C1

Specifications

Melting Point 19°C to 20°C
Boiling Point 234°C
Quantity 25 g
Odor Stench
UN Number UN2810
Beilstein 606496
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzyl mercaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.