missing translation for 'onlineSavingsMsg'
Learn More

4'-Chlorobiphenyl-4-carbaldehyde, TRC

Product Code. 30361231
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361231 5 g 5g
30349389 1 g -
30359212 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361231 Supplier Toronto Research Chemicals Supplier No. TRCC3600205G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80565-30-6
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
Synonym 4'-Chloro[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Chlorophenyl)benzaldehyde, 4'-Chloro-1,1'-biphenyl-4-carboxaldehyde, 4'-Chlorobiphenyl-4-carboxaldehyde
IUPAC Name 4-(4-chlorophenyl)benzaldehyde
SMILES Clc1ccc(cc1)c2ccc(C=O)cc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H9ClO/c14-13-7-5-12(6-8-13)11-3-1-10(9-15)2-4-11/h1-9H
Formula Weight 216.0342
Chemical Name or Material 4'-Chlorobiphenyl-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.