missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyroyl Chloride, TRC

Product Code. 30362362
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362362 50 g 50g
30348832 10 g 10g
30360020 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362362 Supplier Toronto Research Chemicals Supplier No. TRCC36482550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4635-59-0
Molecular Formula C4H6Cl2O
Molecular Weight (g/mol) 140.99
Synonym 4-Chloro-Butanoyl Chloride, 4-Chlorobutanoic Acid Chloride, 4-Chlorobutanoyl Chloride, 4-Chlorobutyric Acid Chloride, 4-Chlorobutyroyl Chloride, 4-Chlorobutyryl Chloride, γ-Chlorobutyroyl Chloride, γ-Chlorobutyryl Chloride, ω-Chlorobutanoyl Chloride, ω-Chlorobutyryl Chloride
IUPAC Name 4-chlorobutanoyl chloride
SMILES ClCCCC(=O)Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H6Cl2O/c5-3-1-2-4(6)7/h1-3H2
Chemical Name or Material 4-Chlorobutyroyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.