missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocarbonylbenzeneboronic anhydride, tech. 90%, Thermo Scientific™

Product Code. 15420156
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420156 1 g 1g
15430156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420156 Supplier Thermo Scientific Alfa Aesar Supplier No. H52852.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332154-57-1
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD06801674
InChI Key ARKQVAQLGLIPAU-UHFFFAOYSA-N
Synonym 4-chlorocarbonylphenylboronic acid, benzoylchloride-4-boronic acid, 4-chlorocarbonyl phenyl boronic acid, boronic acid, b-4-chlorocarbonyl phenyl, 4-chlorocarbonylphenyl boronic anhydride, p-chlorocarbonylphenylboronic acid, 4-chlorocarbonylphenylboronic boronic acid, 4-carbonochloridoyl phenylboronic acid, pubchem9545
PubChem CID 15345913
IUPAC Name (4-carbonochloridoylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)Cl)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 166.37
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Chlorocarbonylbenzeneboronic anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.