missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorodehydromethyltestosterone, TRC

Product Code. 30363491
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363491 5 mg 5mg
30349550 10 mg 10mg
30359378 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363491 Supplier Toronto Research Chemicals Supplier No. TRCC3652205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydrochloromethyltestosterone
CAS 2446-23-3
Purity Grade Notes HPLC
Molecular Formula C20 H27 Cl O2
Quantity 5 mg
Synonym Androsta-1,4-dien-3-one, 4-chloro-17-hydroxy-17-methyl-, (17β)-, Androsta-1,4-dien-3-one, 4-chloro-17β-hydroxy-17-methyl- (7CI,8CI), (17β)-4-Chloro-17-hydroxy-17-methylandrosta-1,4-dien-3-one, 1-Dehydro-4-chloro-17-methyltestosterone, 1-Dehydro-4-chloro-17α-methyltestosterone, 4-Chloro-1,2-dehydro-17α-methyltestosterone, 4-Chloro-1-dehydro-17-methyltestosterone, 4-Chloro-1-dehydromethyltestosterone, 4-Chloro-17α-methyl-17β-hydroxyandrosta-1,4-dien-3-one, 4-Chloro-17β-hydroxy-17α-methyl-5α-androsta-1,4-dien-3-one, 4-Chlorodianabol, 4-Chloromethandienone, Oral-Turinabol
InChI Formula InChI=1S/C20H27ClO2/c1-18-9-8-16(22)17(21)15(18)5-4-12-13(18)6-10-19(2)14(12)7-11-20(19,3)23/h8-9,12-14,23H,4-7,10-11H2,1-3H3/t12-,13+,14+,18-,19+,20+/m1/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CCC4=C(Cl)C(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8R,9S,10R,13S,14S,17S)-4-chloro-17-hydroxy-10,13,17-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 334.88
Formula Weight 334.17
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.