missing translation for 'onlineSavingsMsg'
Learn More

4-(chloromethyl)-2-(2-thienyl)-1,3-thiazole, 97%, Thermo Scientific™

Product Code. 10663783
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10663783 250 mg 250mg
10110592 1 g 1g
10341705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663783 Supplier Thermo Scientific Maybridge Supplier No. CC11524CB
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 54679-16-2
Molecular Formula C8H6ClNS2
Molecular Weight (g/mol) 215.71
MDL Number MFCD01571296
InChI Key CGJJBHKYGNSTDK-UHFFFAOYSA-N
PubChem CID 736496
IUPAC Name 4-(chloromethyl)-2-thiophen-2-yl-1,3-thiazole
SMILES ClCC1=CSC(=N1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 4-chloromethyl-2-2-thienyl-1,3-thiazole, thiazole, 4-chloromethyl-2-2-thienyl, 4-chloromethyl-2-thiophen-2-yl-1,3-thiazole, 4-chloromethyl-2-thiophen-2-yl thiazole, thiazole,4-chloromethyl-2-2-thienyl, 4-chloromethyl-2-thien-2-yl-1,3-thiazole, 4-chloromethyl-2-2-thienyl thiazole, 2-2-thienyl-4-chloromethyl thiazole
Formula Weight 215.73
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)-2-(2-thienyl)-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.