missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride, 97+%, Thermo Scientific™

Product Code. 10249782
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10249782 250 mg 250mg
10259782 1 g 1g
10396374 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249782 Supplier Thermo Scientific Maybridge Supplier No. MO01135CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-83-8
Molecular Formula C10H8Cl3NS
Molecular Weight (g/mol) 280.59
MDL Number MFCD05664422
InChI Key CSXLYGPSLQHDEK-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-3-chlorophenyl thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole-hydrogen chloride 1/1
PubChem CID 2794670
IUPAC Name 4-(chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole;hydrochloride
SMILES Cl.ClCC1=CSC(=N1)C1=CC(Cl)=CC=C1

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 280.61
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.