missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-2-methylthiazole hydrochloride, 98%

Product Code. 11472707
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472707 5 g 5g
11462707 1 g 1g
11482707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472707 Supplier Thermo Scientific Alfa Aesar Supplier No. A14902.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 77470-53-2 | C5H7Cl2NS | 184.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77470-53-2
Molecular Formula C5H7Cl2NS
Molecular Weight (g/mol) 184.08
MDL Number MFCD00067725
InChI Key YGKDISJLDVGNOR-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
PubChem CID 2734203
SMILES Cl.CC1=NC(CCl)=CS1

Specifications

Melting Point ∼165°C (decomposition)
Quantity 5 g
UN Number UN3261
Beilstein 108603
IUPAC Name 4-(chloromethyl)-2-methyl-1,3-thiazole hydrochloride
Formula Weight 184.09
Percent Purity 98%
Chemical Name or Material 4-Chloromethyl-2-methylthiazole hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.