missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole, 97%

Product Code. 15452068
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452068 5 g 5g
15472256 250 mg 250mg
15442068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452068 Supplier Thermo Scientific Alfa Aesar Supplier No. H55735.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80841-78-7 | C5H5ClO3 | 148.542 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80841-78-7
Molecular Formula C5H5ClO3
Molecular Weight (g/mol) 148.542
MDL Number MFCD07787494
InChI Key QCLFSYYUWPUWQR-UHFFFAOYSA-N
Synonym 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, pubchem10542, acmc-209pk7, ksc490e4r, 4-chloromethyl-5methyl-1,3-dioxol-2-one
PubChem CID 9855518
IUPAC Name 4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
SMILES CC1=C(OC(=O)O1)CCl

Specifications

Boiling Point >90°C (2 mmHg)
Flash Point 129°C (264°F)
Quantity 5 g
Beilstein 4798926
Formula Weight 148.55
Percent Purity 97%
Chemical Name or Material 4-Chloromethyl-5-methyl-2-oxo-1,3-dioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.