missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)phenyltrichlorosilane, 97%

Product Code. p-7046812
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11389155

Brand: Thermo Scientific Alfa Aesar L16431.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13688-90-9 | C7H6Cl4Si | 260.01 g/mol

4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13688-90-9
Molecular Formula C7H6Cl4Si
Molecular Weight (g/mol) 260.01
MDL Number MFCD00045268
InChI Key YBYBQFPZMKPGPJ-UHFFFAOYSA-N
Synonym 4-chloromethyl phenyltrichlorosilane, p-chloromethyl phenyltrichlorosilane, silane, trichloro 4-chloromethyl phenyl, trichloro 4-chloromethyl phenyl silane, silane, trichloro .alpha.-chloro-p-tolyl, benzene, 1-chloromethyl-4-trichlorosilyl, p-chloromethylphenyltrichlorosilane
PubChem CID 83663
IUPAC Name trichloro-[4-(chloromethyl)phenyl]silane
SMILES C1=CC(=CC=C1CCl)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 142°C to 144°C (15mmHg)
Quantity 5 g
UN Number UN2987
Beilstein 3090295
Flash Point 78°C (172°F)
Solubility Information Reacts with water.
Formula Weight 260.02
Percent Purity 97%
Refractive Index 1.548
Sensitivity Moisture sensitive; May form a precipitate
Density 1.361
Chemical Name or Material 4-(Chloromethyl)phenyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.