missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)pyridine Hydrochloride, TRC
Click to view available options
Quantity:
10 g
Specifications
Specifications
| Chemical Name or Material | 4-(Chloromethyl)pyridine Hydrochloride |
| CAS | 1822-51-1 |
| Molecular Formula | C6H7Cl2N |
| Quantity | 10 g |
| Synonym | 4-(Chloromethyl)pyridine Hydrochloride, 4-(Chloromethyl)pyridinium Chloride, 4-Chloromethylpyridine hydrochloride Salt, 4-Picolyl Chloride Hydrochloride, 4-Pyridylmethyl Chloride Hydrochloride |
| InChI Formula | InChI=1S/C6H6ClN.ClH/c7-5-6-1-3-8-4-2-6;/h1-4H,5H2;1H |
| SMILES | Cl.ClCc1ccncc1 |
| IUPAC Name | 4-(chloromethyl)pyridine;hydrochloride |
| Molecular Weight (g/mol) | 164.03 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction