missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenethyl bromide, 95%

Product Code. 11944541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11944541 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11944541 Supplier Thermo Scientific Acros Supplier No. 440180050
 Restricted Item
Estimated Shipment: 10-08-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6529-53-9 | C8H8BrCl | 219.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6529-53-9
CAS Min % 94.0
Color Colorless to Yellow
CAS Max % 100.0
Density 1.5080g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 94% min. (GC)
Specific Gravity 1.508
Molecular Formula C8H8BrCl
Synonym 1-2-bromoethyl-4-chlorobenzene, 4-chlorophenethyl bromide, 2-4-chlorophenyl ethylbromide, 1-bromo-2-4-chlorophenyl ethane, benzene, 1-2-bromoethyl-4-chloro, p-chlorophenethyl bromide, 4-chlorophenethyl bromide 97, p-chloro-.beta.-phenethyl bromide, 1-chloro-4-2-bromoethyl benzene, acmc-20aa8v
Boiling Point 85°C (1.0 Torr)
InChI Key YAFMYKFAUNCQPU-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCBr)Cl
IUPAC Name 1-(2-bromoethyl)-4-chlorobenzene
Molecular Weight (g/mol) 219.51
PubChem CID 23029
Formula Weight 219.51
Percent Purity 95%
Chemical Name or Material 4-Chlorophenethyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chlorophenethyl bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.