missing translation for 'onlineSavingsMsg'
Learn More

4-chlorophenoxyacetonitrile, 98%, ACROS Organics™

Product Code. 10833971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10833971 5 g 5g
10832631 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10833971 Supplier Thermo Scientific Acros Supplier No. 437600050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3598-13-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00031535
InChI Key YUGDKEWUYZXXRU-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
PubChem CID 306444
IUPAC Name 2-(4-chlorophenoxy)acetonitrile
SMILES ClC1=CC=C(OCC#N)C=C1

Specifications

Melting Point 45°C to 49°C
CAS Min % 97.5
Color Tan-Yellow
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Physical Form Powder
Assay Percent Range 98%
Linear Formula ClC6H4OCH2CN
Formula Weight 167.6
Percent Purity ≥97.5%
Chemical Name or Material 4-chlorophenoxyacetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.