missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)-4-piperidinylmethanone Hydrochloride, TRC

Product Code. 30367785
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30367785 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30367785

Brand: Toronto Research Chemicals TRCC379585500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Chlorophenyl)-4-piperidinyl-methanone, Hydrochloride
CAS 55695-51-7
Molecular Formula C12 H14 Cl N O . Cl H
Quantity 500 mg
InChI Formula InChI=1S/C12H14ClNO.ClH/c13-11-3-1-9(2-4-11)12(15)10-5-7-14-8-6-10;/h1-4,10,14H,5-8H2;1H
SMILES Cl.Clc1ccc(cc1)C(=O)C2CCNCC2
IUPAC Name (4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride
Molecular Weight (g/mol) 260.16
Formula Weight 259.0531
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.