missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl cyclopropyl ketone, 98%

Product Code. 11439797
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439797 250 g 250g
11419797 10 g 10g
11429797 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439797 Supplier Thermo Scientific Alfa Aesar Supplier No. B22766.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6640-25-1 | C10H9ClO | 180.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6640-25-1
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00001295
InChI Key OPSFCTBBDIDFJM-UHFFFAOYSA-N
Synonym 4-chlorophenyl cyclopropyl ketone, 4-chlorophenyl cyclopropyl methanone, 4-chlorobenzoyl cyclopropane, 4-chlorobenzoylcyclopropane, methanone, 4-chlorophenyl cyclopropyl, 4-chlorophenylcyclopropylmethanone, pubchem16774, acmc-209nvc, cyclopropyl 4-chlorophenyl ketone
PubChem CID 81148
IUPAC Name (4-chlorophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 29°C to 31°C
Density 1.16
Boiling Point 135°C to 137°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5718
Quantity 250 g
Beilstein 2044845
Formula Weight 180.63
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl cyclopropyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.