missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetyl chloride, 98%

Product Code. 11474194
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474194 250 g 250g
11454194 10 g 10g
11464194 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474194 Supplier Thermo Scientific Alfa Aesar Supplier No. A12530.30
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25026-34-0
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00037111
InChI Key UMQUIRYNOVNYPA-UHFFFAOYSA-N
Synonym 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
PubChem CID 90692
IUPAC Name 2-(4-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=C(Cl)C=C1

Specifications

Density 1.292
Quantity 250 g
Refractive Index 1.551
UN Number UN3265
Beilstein 637775
Boiling Point 112°C to 114°C (11 mmHg)
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.