missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals

Product Code. 15438047
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438047 100 mL 100mL
15448047 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438047 Supplier Thermo Scientific Alfa Aesar Supplier No. H51163.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-77-8
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00009926
InChI Key CDEMHJCJMMOFMB-UHFFFAOYSA-M
Synonym 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent
PubChem CID 101888
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC=C([Mg]Br)C=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 215.76
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 4-Chlorophenylmagnesium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.