missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloropropiophenone, 97%, Thermo Scientific™

Product Code. p-91603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10734601 25 g Glass bottle 25g
10734791 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734601 Supplier Thermo Scientific Acros Supplier No. 110000250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6285-05-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000626
InChI Key ADCYRBXQAJXJTD-UHFFFAOYSA-N
Synonym 4'-chloropropiophenone, 1-4-chlorophenyl propan-1-one, p-chloropropiophenone, 4-chloropropiophenone, 1-propanone, 1-4-chlorophenyl, propiophenone, 4'-chloro, propiophenone, p-chloro, ethyl p-chlorophenyl ketone, usaf ek-5296, unii-26ay8wja1e
PubChem CID 22697
IUPAC Name 1-(4-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H4COC2H5
Quantity 25 g
Beilstein 07, 301
Formula Weight 168.62
Percent Purity ≥96.0%
Physical Form Crystalline Mass
Chemical Name or Material 4';-Chloropropiophenone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.