missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroquinoline-6-boronic acid pinacol ester, 96%

Product Code. 15450746
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450746 250 mg 250mg
15440746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450746 Supplier Thermo Scientific Alfa Aesar Supplier No. H54085.MD
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1201844-73-6 | C15H17BClNO2 | 289.566 g/mol

4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1201844-73-6
Molecular Formula C15H17BClNO2
Molecular Weight (g/mol) 289.566
MDL Number MFCD16987803
InChI Key ANPKKHFSNJKOAM-UHFFFAOYSA-N
Synonym 4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 4-chloroquinoline-6-boronic acid pinacol ester, quinoline, 4-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 53363436
IUPAC Name 4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=CN=C3C=C2)Cl

Specifications

Quantity 250 mg
Formula Weight 289.56
Percent Purity 96%
Chemical Name or Material 4-Chloroquinoline-6-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.