missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorotoluene-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorotoluene-2,3,5,6-D4 |
| CAS | 85577-24-8 |
| Molecular Formula | C7 D4 H3 Cl |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,4,5-d4, 3-chloro-6-methyl- (9 CI), 1-Chloro-4-methyl-2,3,5,6-tetradeuterobenzene, 3-Chloro-6-methylbenzene-1,2,4,5-d4, 4-Chlorotoluene-d4, 4-Chlorotoluene-2,3,5,6-d4, 4-Chlorotoluene-2,3,5,6-D4 |
| InChI Formula | InChI=1 S/C7H7Cl/c1-6-2-4-7(8)5-3-6/h2-5 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c([2 H])c([2 H])c1C |
| IUPAC Name | 1-chloro-2,3,5,6-tetradeuterio-4-methylbenzene |
| Molecular Weight (g/mol) | 130.6081 |
| Formula Weight | 130.0487 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction