missing translation for 'onlineSavingsMsg'
Learn More

4-chlorotoluene, 98%

Product Code. 10086003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10086003 100 mL Glass bottle 100mL
10562161 5 mL Glass bottle 5mL
10229313 25 mL Glass bottle 25mL
10306460 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10086003 Supplier Thermo Scientific Acros Supplier No. 150211000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-43-4 | C7H7Cl | 126.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-43-4
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000631
InChI Key NPDACUSDTOMAMK-UHFFFAOYSA-N
Synonym 4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n
PubChem CID 7810
ChEBI CHEBI:34401
IUPAC Name 1-chloro-4-methylbenzene
SMILES CC1=CC=C(Cl)C=C1

Specifications

Melting Point 6.0°C to 8.0°C
CAS Min % 97.5
CAS Max % 100.0
Density 1.0700g/mL
Quantity 100 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5190 to 1.5210
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.07
Linear Formula CH3C6H4Cl
Beilstein 05, 292
Merck Index 15, 2175
Boiling Point 162.0°C
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,chloroform and ether
Formula Weight 126.59
Percent Purity 98%
Chemical Name or Material 4-Chlorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.