missing translation for 'onlineSavingsMsg'
Learn More

4-Cholesten-3-one, 98+%

Product Code. 11449283
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449283 5 g 5g
11459283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11743.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 601-57-0 | C27H44O | 384.648 g/mol

4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cholesten-3-one is used as a chiral building block for proteomics research. It is an intestinal metabolite of cholesterol and exhibits an anti-obesity effect on animals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601-57-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
MDL Number MFCD00003663
InChI Key NYOXRYYXRWJDKP-GYKMGIIDSA-N
Synonym 4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4
PubChem CID 91477
ChEBI CHEBI:16175
IUPAC Name (8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C

Specifications

Melting Point 79°C to 81°C
Quantity 5 g
Beilstein 4707774
Solubility Information Slightly soluble in water.
Optical Rotation +93° (c=1 in chloroform)
Formula Weight 384.65
Percent Purity ≥98%
Chemical Name or Material 4-Cholesten-3-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.