missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanol, 97%

Product Code. 11422777
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422777 5 g 5g
11412777 1 g 1g
11432777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422777 Supplier Thermo Scientific Alfa Aesar Supplier No. A14940.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1481-93-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1481-93-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00006849
InChI Key MGSHXMOLUWTMGP-UHFFFAOYSA-N
PubChem CID 92890
IUPAC Name 3,4-dihydro-2H-chromen-4-ol
SMILES C1COC2=CC=CC=C2C1O

Specifications

Melting Point 38°C to 42°C
Boiling Point 79°C to 81°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 130800
Synonym 4-chromanol, chroman-4-ol, 3,4-dihydro-2h-1-benzopyran-4-ol, 4-hydroxychroman, 2h-1-benzopyran-4-ol, 3,4-dihydro, acmc-20mukj, 3'-carboxy-alpha-chromanol, 2h-1-benzopyran-4-ol,3,4-dihydro
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 4-Chromanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.