missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorophenylboronic acid, 97%

Product Code. 11906141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11906141 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11906141 Supplier Thermo Scientific Acros Supplier No. 445390010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1150114-77-4 | C7H5BFNO2 | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1150114-77-4
Melting Point 304°C to 304.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H5BFNO2
MDL Number MFCD07644472
Quantity 1 g
Synonym 4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid
InChI Key HFAUMAIVCYTVQR-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C=C(C=C1)C#N
IUPAC Name (4-cyano-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 164.93
PubChem CID 44755209
Formula Weight 164.93
Percent Purity 97%
Chemical Name or Material 4-Cyano-2-fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyano-2-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.