missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluoropyridine, 97%

Product Code. 11485700
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11485700

Brand: Thermo Scientific Alfa Aesar H25938.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3939-14-8 | C6H3FN2 | 122.102 g/mol

4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluoropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride, nitrogen oxides, strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3939-14-8
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD09027299
InChI Key DYQMGPSBUBTNJZ-UHFFFAOYSA-N
PubChem CID 12002587
IUPAC Name 2-fluoropyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)F

Specifications

Melting Point 31°C to 33°C
Quantity 1 g
Synonym 2-fluoroisonicotinonitrile, 4-cyano-2-fluoropyridine, 2-fluoro-4-cyanopyridine, 4-pyridinecarbonitrile, 2-fluoro, 2-fluoro-4-pyridinecarbonitrile, 2-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,2-fluoro, acmc-1bnd8, 4-cyano-2-fluoro pyridine, ksc581g3h
Solubility Information Insoluble in water.
Formula Weight 122.1
Percent Purity 97%
Chemical Name or Material 4-Cyano-2-fluoropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.