missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-fluorophenylboronic acid, 97%

Product Code. 13498608 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13498608 5 g 5g
13488608 1 g 1g
13408618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13498608 Supplier Thermo Scientific Acros Supplier No. 448710050
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 843663-18-3 | C7H5BFNO2 | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 843663-18-3
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03411549
InChI Key DECWLXUOZUMPBF-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug
PubChem CID 2783130
IUPAC Name (4-cyano-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(C=C1)C#N

Specifications

Melting Point 362°C to 366°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.93
Percent Purity 97%
Chemical Name or Material 4-Cyano-3-fluorophenylboronic acid
compliance-icons
Product Identifier
  • 4-Cyano-3-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.