missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10663645
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10663645 25 g 25g
10243712 1 g 1g
10559972 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663645 Supplier Thermo Scientific Maybridge Supplier No. TL00421EB
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 49584-26-1
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.62
MDL Number MFCD00052337
InChI Key DBMFYTQPPBBKHI-UHFFFAOYSA-N
Synonym 4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
PubChem CID 39497
IUPAC Name 4-cyanobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CNC6H4SO2Cl
Quantity 25 g
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.