missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzeneboronic acid, 98%

Product Code. 11374865
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11374865 1 g 1g
11384865 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11374865

Brand: Thermo Scientific Alfa Aesar L18007.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126747-14-6 | C7H6BNO2 | 146.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126747-14-6
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318968
InChI Key CEBAHYWORUOILU-UHFFFAOYSA-N
Synonym 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
PubChem CID 2734326
IUPAC Name (4-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C#N

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 6593772
Formula Weight 146.94
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.