missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyanomethoxy)phenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11916141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11916141 1 g 1g
11926141 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916141 Supplier Thermo Scientific Acros Supplier No. 445400010
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 947533-23-5 | Molecular Formula: C8H8BNO3 | Molecular Weight: 176.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947533-23-5
Molecular Formula C8H8BNO3
Molecular Weight (g/mol) 176.97
MDL Number MFCD09908170
InChI Key GKJPNYHKHSLHCC-UHFFFAOYSA-N
Synonym 4-cyanomethoxyphenylboronic acid, 4-cyanomethoxy phenyl boronic acid, 4-cyanomethoxy phenylboronic acid, acmc-209rtj
PubChem CID 21918429
IUPAC Name [4-(cyanomethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OCC#N)C=C1

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 176.97
Percent Purity 97%
Chemical Name or Material 4-(Cyanomethoxy)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(Cyanomethoxy)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.