missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclopropyl-4-oxobutyric acid, 95%, Thermo Scientific Chemicals

Product Code. 15497957
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15497957 Supplier Thermo Scientific Alfa Aesar Supplier No. H51078.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53712-75-7
Molecular Formula C7H10O3
MDL Number MFCD01320151
Quantity 1 g
Synonym 4-cyclopropyl-4-oxobutyric acid, acmc-1ayb0, 4-oxo-4-cyclopropylbutanoic acid
InChI Key ISZLFKWEBSKHTO-UHFFFAOYSA-N
SMILES OC(=O)CCC(=O)C1CC1
IUPAC Name 4-cyclopropyl-4-oxobutanoic acid
Molecular Weight (g/mol) 142.15
PubChem CID 16213729
Formula Weight 142.15
Percent Purity 95%
Chemical Name or Material 4-Cyclopropyl-4-oxobutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.