missing translation for 'onlineSavingsMsg'
Learn More

4-Decanolide, TRC

Product Code. 30362405
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362405 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362405 Supplier Toronto Research Chemicals Supplier No. TRCD21255350G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material gamma-Decalactone
CAS 706-14-9
Molecular Formula C10 H18 O2
Quantity 50 g
Synonym 2-Decalactone, 4-Hexyl-γ-butyrolactone, γ-Hexyl-γ-butyrolactone, 5-Hexyltetrahydro-2-furanone, Decanolactone, 4-Hydroxydecanoic acid γ-lactone, γ-Decanolactone
InChI Formula InChI=1S/C10H18O2/c1-2-3-4-5-6-9-7-8-10(11)12-9/h9H,2-8H2,1H3
SMILES O=C1OC(CC1)CCCCCC
IUPAC Name 5-hexyltetrahydrofuran-2-one
Molecular Weight (g/mol) 170.25
Formula Weight 170.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.