missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 4'-Demethylepipodophyllotoxin

Product Code. 16457306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457306 Supplier MedChemExpress Supplier No. HY17435
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 4'-Demethylepipodophyllotoxin(4'-DMEP) is a key intermediate compound for the preparation of podophyllotoxin-type anti-cancer drugs; a potent inhibitor of microtubule assembly.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 400.38 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 6559-91-7
Product Type Natural Products
Purity 99.75%
Solubility DMSO : 100 mg/mL (249.76 mM∣Need ultrasonic)
Synonym 4'-O-demethylepipodophyllotoxin∣ 4'-DMEP
Target Microtubule/Tubulin
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.