missing translation for 'onlineSavingsMsg'
Learn More

4-Deoxypodophyllotoxin, TRC

Product Code. 30137497
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137497 1 g 1g
18092740 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137497 Supplier Toronto Research Chemicals Supplier No. TRCD2495001G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19186-35-7
Molecular Formula C22 H22 O7
Molecular Weight (g/mol) 398.41
Synonym AS 2-3, Anthricin, Deoxypodophyllotoxin, Desoxypodophyllotoxin, Silicicolin, (-)-Anthricin, (-)-Deoxypodophyllotoxin, (-)-Desoxypodophyllotoxin;
IUPAC Name (5R,5aR,8aR)-5-(3,4,5-trimethoxyphenyl)-5a,8,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-6-one
SMILES COc1cc(cc(OC)c1OC)[C@H]2[C@@H]3[C@H](COC3=O)Cc4cc5OCOc5cc24

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H22O7/c1-24-17-6-12(7-18(25-2)21(17)26-3)19-14-8-16-15(28-10-29-16)5-11(14)4-13-9-27-22(23)20(13)19/h5-8,13,19-20H,4,9-10H2,1-3H3/t13-,19+,20-/m0/s1
Formula Weight 398.1366
Percent Purity >95%
Chemical Name or Material 4-Deoxypodophyllotoxin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.