missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethoxy-4-chloro Omeprazole, TRC

Product Code. 30366485
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366485 1 mg 1mg
30361273 10 mg 10mg
30365800 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366485 Supplier Toronto Research Chemicals Supplier No. TRCD2903901MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863029-89-4
Molecular Formula C16 H16 Cl N3 O2 S
Molecular Weight (g/mol) 349.84
Synonym 2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, Omeprazole Imp. H (EP)
IUPAC Name 2-[(4-chloro-3,5-dimethylpyridin-2-yl)methylsulfinyl]-5-methoxy-1H-benzimidazole
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(Cl)c3C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H16ClN3O2S/c1-9-7-18-14(10(2)15(9)17)8-23(21)16-19-12-5-4-11(22-3)6-13(12)20-16/h4-7H,8H2,1-3H3,(H,19,20)
Formula Weight 349.0652
Chemical Name or Material 2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.