missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethoxy Omeprazole, TRC

Product Code. 30007451
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007451 50 mg 50mg
30004779 5 mg 5mg
30007438 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007451 Supplier Toronto Research Chemicals Supplier No. TRCD29056050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110374-16-8
Molecular Formula C16 H17 N3 O2 S
Molecular Weight (g/mol) 315.39
Synonym 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Esomeprazole Magnesium Trihydrate Imp. B (EP), Omeprazole Imp. B (EP)
IUPAC Name 2-[(3,5-dimethylpyridin-2-yl)methylsulfinyl]-5-methoxy-1H-benzimidazole
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)cc3C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H17N3O2S/c1-10-6-11(2)15(17-8-10)9-22(20)16-18-13-5-4-12(21-3)7-14(13)19-16/h4-8H,9H2,1-3H3,(H,18,19)
Formula Weight 315.1041
Chemical Name or Material 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.