missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethoxypropoxyl-4-methoxy Rabeprazole, TRC

Product Code. 30011249
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30011249 100 mg 100mg
30006076 10 mg 10mg
30005007 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30011249 Fournisseur Toronto Research Chemicals Code fournisseur TRCD290610100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 102804-77-3
Molecular Formula C15 H15 N3 O2 S
Molecular Weight (g/mol) 301.36
Synonym 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Rabeprazole Sodium Imp E (EP)
IUPAC Name 2-[(4-methoxy-3-methylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES COc1ccnc(CS(=O)c2nc3ccccc3[nH]2)c1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H15N3O2S/c1-10-13(16-8-7-14(10)20-2)9-21(19)15-17-11-5-3-4-6-12(11)18-15/h3-8H,9H2,1-2H3,(H,17,18)
Formula Weight 301.0885
Percent Purity >95%
Chemical Name or Material 4-Desmethoxypropoxyl-4-methoxy Rabeprazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.