missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)azobenzene, 98%

Product Code. 11426776
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426776 5 g 5g
11436776 25 g 25g
11446776 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426776 Supplier Thermo Scientific Alfa Aesar Supplier No. B23915.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2481-94-9 | C16H19N3 | 253.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2481-94-9
Molecular Formula C16H19N3
Molecular Weight (g/mol) 253.349
MDL Number MFCD00009043
InChI Key SJJISKLXUJVZOA-UHFFFAOYSA-N
Synonym 4-diethylamino azobenzene, oil yellow de, diethyl yellow, c.i. solvent yellow 56, solvent yellow 56, 4-diethylaminoazobenzene, oil yellow ga, oil yellow nb, oil yellow dea, oil yellow enc
PubChem CID 17204
IUPAC Name N,N-diethyl-4-phenyldiazenylaniline
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=CC=C2

Specifications

Melting Point 93°C to 96°C
Quantity 5 g
Formula Weight 253.35
Percent Purity 98%
Chemical Name or Material 4-(Diethylamino)azobenzene
compliance-icons
Product Identifier
  • 4-Diethylaminoazobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N-diethyl-4-(phenylazo)Benzenamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.